A14929.09
A14929.18
A14929.30
A14929.09EA
56.3
EUR
InStock
A14929.09
A14929.18
A14929.30
Tetraoctylammoniumbromid ≥98%
Tetraoctylammoniumbromid
Purity: 98+%
CAS No.: 14866-33-2
Molecular Formula: [CH3(CH2)7]4NBr
Formula Weight: 546.81
Melting Point: 94-99°
Beilstein Reference: 4169176