A14936.03
A14936.06
A14936.14
A14936.03EA
25.9
EUR
InStock
A14936.03
A14936.06
A14936.14
2,6-Dimethoxy-p-benzochinon ≥98%
2,6-Dimethoxy-p-benzochinon
Purity: 98%
CAS No.: 530-55-2
Molecular Formula: C8H8O4
Formula Weight: 168.15
Melting Point: 252-257°
Beilstein Reference: 2048182
Merck Reference: 14,3225
MDL No.: MFCD00016368