A15199.0E
A15199.18
A15199.30
A15199.36
A15199.0EEA
190
EUR
InStock
A15199.0E
A15199.18
A15199.30
A15199.36
3,4-Dichloranilin ≥98%
3,4-Dichloranilin
Purity: 98%
CAS No.: 95-76-1
Molecular Formula: C6H5Cl2N
Formula Weight: 162.02
Flash Point: 135°(275°F)
Melting Point: 70-73°
Boiling Point: 272°
Beilstein Reference: 636837
Merck Reference: 14,3054
UN No.: 3442
MDL No.: MFCD00007768