A15225.09
A15225.18
A15225.30
A15225.09EA
57.8
EUR
InStock
A15225.09
A15225.18
A15225.30
Benzofuran ≥99%
Benzofuran
Purity: 99+%
CAS No.: 271-89-6
Molecular Formula: C8H6O
Formula Weight: 118.14
Storage Temperature: 2°C to 8°C
Flash Point: 56°(133°F)
Refractive Index: 1.5660
Odour: Aromatic
Boiling Point: 173-175°
Beilstein Reference: 107704
Merck Reference: 14,1088
UN No.: 1993
MDL No.: MFCD00005847