A15229.06
A15229.14
A15229.06EA
69.6
EUR
InStock
A15229.06
A15229.14
3-Cyclohexen-1-carbonsäure 98%
3-Cyclohexen-1-carbonsäure
Purity: 98%
CAS No.: 4771-80-6
Molecular Formula: C7H10O2
Formula Weight: 126.16
Flash Point: 110°(230°F)
Refractive Index: 1.4815
Melting Point: 16-18°
Boiling Point: 120-122°/10mm
Solubility: V sol
Beilstein Reference: 1617723
UN No.: UN3265
MDL No.: MFCD00013781