A15240.22
A15240.36
A15240.22EA
56.3
EUR
InStock
A15240.22
A15240.36
Octamethylcyclotetrasiloxan ≥98%
Octamethylcyclotetrasiloxan
Purity: 98%
CAS No.: 556-67-2
Molecular Formula: C8H24O4Si4
Formula Weight: 296.62
Flash Point: 56°(132°F)
Refractive Index: 1.3960
Melting Point: 17-18°
Boiling Point: 175-176°
Beilstein Reference: 1787074
Merck Reference: 14,6747
UN No.: 1993
MDL No.: MFCD00003269