A15244.06
A15244.14
A15244.06EA
52.2
EUR
InStock
A15244.06
A15244.14
α-Brom-m-toluonitril ≥98%
α-Brom-m-toluonitril
Purity: 98%
CAS No.: 28188-41-2
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Melting Point: 92-96°
Beilstein Reference: 742356
UN No.: 2923
MDL No.: MFCD00001809