A15325.14
A15325.22
A15325.14EA
67.5
EUR
InStock
A15325.14
A15325.22
(4-Bromphenyl)essigsäure ≥99%
(4-Bromphenyl)essigsäure
Purity: 99%
CAS No.: 1878-68-8
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 115-119°
Beilstein Reference: 972146