A15329.06
A15329.14
A15329.22
A15329.06EA
43.2
EUR
InStock
A15329.06
A15329.14
A15329.22
3-Hydroxy-4-nitrobenzoesäuremethylester ≥98%
Methyl-3-hydroxy-4-nitrobenzoate
Purity: 98%
CAS No.: 713-52-0
Molecular Formula: C8H7NO5
Formula Weight: 197.15
Melting Point: 89-92°
Beilstein Reference: 2583832