A15353.06
A15353.14
A15353.22
A15353.06EA
29.7
EUR
InStock
A15353.06
A15353.14
A15353.22
2-Acetyl-1-methylpyrrol ≥98%
2-Acetyl-1-methylpyrrol
Purity: 98%
CAS No.: 932-16-1
Molecular Formula: C7H9NO
Formula Weight: 123.16
Flash Point: 68°(154°F)
Refractive Index: 1.5420
Odour: Pleasant
Boiling Point: 200-202°
Beilstein Reference: 111887
MDL No.: MFCD00003089