A15383.14
A15383.22
A15383.36
A15383.14EA
32.5
EUR
InStock
A15383.14
A15383.22
A15383.36
4-Fluorbenzaldehyd ≥98%
4-Fluorbenzaldehyd
Purity: 98%
CAS No.: 459-57-4
Molecular Formula: C7H5FO
Formula Weight: 124.11
Flash Point: 56°(133°F)
Refractive Index: 1.5210
Melting Point: -10°
Boiling Point: 180-181°
Beilstein Reference: 385857
UN No.: 1989
MDL No.: MFCD00003378