A15435.22
A15435.14
A15435.36
A15435.22EA
138
EUR
InStock
A15435.22
A15435.14
A15435.36
2-Cyclohexen-1-on ≥97%
2-Cyclohexen-1-on
Purity: 97%
CAS No.: 930-68-7
Molecular Formula: C6H8O
Formula Weight: 96.13
Flash Point: 56°(132°F)
Refractive Index: 1.4880
Melting Point: -53°
Boiling Point: 167-169°
Beilstein Reference: 1280477
UN No.: 2929
MDL No.: MFCD00001577