A15458.14
A15458.03
A15458.22
A15458.14EA
215
EUR
InStock
A15458.14
A15458.03
A15458.22
m-Toluolsulfonylchlorid ≥98%
m-Toluolsulfonylchlorid
Purity: 98%
CAS No.: 1899-93-0
Molecular Formula: C7H7ClO2S
Formula Weight: 190.65
Flash Point: 107°(224°F)
Refractive Index: 1.5480
Odour: Acrid
Boiling Point: 252-253°
Beilstein Reference: 2087865
UN No.: 3265
MDL No.: MFCD00051747