A15555.0B
A15555.22
A15555.30
A15555.36
A15555.0BEA
116
EUR
InStock
A15555.0B
A15555.22
A15555.30
A15555.36
Methylcyanacetat ≥99%
Methylcyanacetat
Purity: 99%
CAS No.: 105-34-0
Molecular Formula: NCCH2CO2CH3
Formula Weight: 99.09
Flash Point: 110°(230°F)
Refractive Index: 1.4185
Odour: Mild
Melting Point: -23°
Boiling Point: 204°
Beilstein Reference: 773945
MDL No.: MFCD00001939