A15597.14
A15597.22
A15597.36
A15597.14EA
22.5
EUR
InStock
A15597.14
A15597.22
A15597.36
4-Fluoranilin ≥99%
4-Fluoranilin
Purity: 99%
CAS No.: 371-40-4
Molecular Formula: C6H6FN
Formula Weight: 111.12
Flash Point: 73°(163°F)
Refractive Index: 1.5400
Odour: Recognizable
Melting Point: -2°
Boiling Point: 186-187°
Beilstein Reference: 742030
Merck Reference: 14,4169
UN No.: 2941
MDL No.: MFCD00007829