A15754.03
A15754.06
A15754.03EA
21.1
EUR
InStock
A15754.03
A15754.06
5-Amino-1-methyl-3-phenyl-1H-pyrazol 97%
5-Amino-1-methyl-3-phenyl-1H-pyrazole
Purity: 97%
CAS No.: 10199-50-5
Molecular Formula: C10H11N3
Formula Weight: 173.22
Odour: Odourless
Melting Point: 124-130°
Beilstein Reference: 743807
MDL No.: MFCD00067874