A15782.14
A15782.22
A15782.36
A15782.14EA
75.9
EUR
InStock
A15782.14
A15782.22
A15782.36
2,2′-Bipyridin ≥99%
2,2'-Bipyridyl
Purity: 99+%
CAS No.: 366-18-7
Molecular Formula: C10H8N2
Formula Weight: 156.19
Flash Point: 121°(249°F)
Melting Point: 70°
Boiling Point: 272-273°
Beilstein Reference: 113089
Merck Reference: 14,3347
UN No.: 2811
MDL No.: MFCD00006212