A15809.30
A15809.0B
A15809.0I
A15809.30EA
28.1
EUR
InStock
A15809.30
A15809.0B
A15809.0I
Brij® 35
Brij® 35
CAS No.: 9002-92-0
Molecular Formula: C58H118O24
Formula Weight: 1199.57
Physical Form: Waxy solid
Appearance: White
Melting Point: ca 35-40°
MDL No.: MFCD00080891
For use in Stein-Moore chromatography