A16084.0J
A16084.AK
A16084.AU
A16084.PI
A16084.0JEA
166
EUR
InStock
A16084.0J
A16084.AK
A16084.AU
A16084.PI
Tributylphosphat ≥98%
Tributylphosphat
Purity: 98%
CAS No.: 126-73-8
Molecular Formula: C12H27O4P
Formula Weight: 266.32
Flash Point: 152°(305°F)
Physical Form: Liquid
Density: 0.977
Refractive Index: 1.4240
Appearance: Colourless
Odour: Odourless
Melting Point: -80°
Boiling Point: 155-156°/10mm
Beilstein Reference: 1710584
Merck Reference: 14,9618
MDL No.: MFCD00009436