A16143.0E
A16143.30
A16143.36
A16143.0EEA
123
EUR
InStock
A16143.0E
A16143.30
A16143.36
(±)-3-Chlor-1,2-propandiol ≥98%
(±)-3-Chlor-1,2-propandiol
Purity: 98%
CAS No.: 96-24-2
Molecular Formula: C3H7ClO2
Formula Weight: 110.54
Flash Point: 137°(278°F)
Refractive Index: 1.4800
Melting Point: -40°
Boiling Point: 213°
Beilstein Reference: 635684
Merck Reference: 14,2145
UN No.: 2689
MDL No.: MFCD00004712