A16203.30
A16203.0B
A16203.0I
A16203.30EA
38.9
EUR
InStock
A16203.30
A16203.0B
A16203.0I
Kupfer(II)acetat Monohydrat ≥98%
Kupfer(II)acetat Monohydrat
Purity: 98+%
CAS No.: 6046-93-1
Molecular Formula: C4H6CuO4.H2O
Formula Weight: 199.65 (181.64anhy)
Physical Form: Powder
Density: 1.882
Appearance: Blue-green
Odour: Odourless
pH: 5.2-5.5
Melting Point: 115°
Merck Reference: 14,2624
UN No.: 3077
MDL No.: MFCD00149570