A16236.0B
A16236.18
A16236.30
A16236.0BEA
193
EUR
InStock
A16236.0B
A16236.18
A16236.30
Bis(4-hydroxy-N-methylanilinium)sulfat 99%
4-Methylaminophenolsulfat
Purity: 99%
CAS No.: 55-55-0
Molecular Formula: C14H18N2O2.H2SO4
Formula Weight: 344.38
Melting Point: ca 260° dec.
Beilstein Reference: 3919382
Merck Reference: 14,6017
UN No.: 2811
MDL No.: MFCD00013140