A16237.14
A16237.22
A16237.36
A16237.14EA
31
EUR
InStock
A16237.14
A16237.22
A16237.36
Vitamin A acetat in Gelatine, 500,000 I.U./g
alle-trans-Retinylacetat
CAS No.: 127-47-9
Beilstein Reference: 1915439
Merck Reference: 14,10013
MDL No.: MFCD00019413