A16502.0B
A16502.30
A16502.0BEA
182
EUR
InStock
A16502.0B
A16502.30
3,5-Dinitrobenzoesäure ≥98%
3,5-Dinitrobenzoic acid
Purity: 98+%
CAS No.: 99-34-3
Molecular Formula: C7H4N2O6
Formula Weight: 212.12
Melting Point: 204-208°
Beilstein Reference: 1914286
Merck Reference: 14,3276
UN No.: 1325
MDL No.: MFCD00007253