A16560.18
A16560.30
A16560.18EA
76.6
EUR
InStock
A16560.18
A16560.30
Chromium(III)-2-ethylhexanoate 50% in 2-Ethylhexansäure
Chromium(III)-2-ethylhexanoate
CAS No.: 3444-17-5
Molecular Formula: C24H45CrO6
Formula Weight: 481.62
Odour: Weak, characteristic
MDL No.: MFCD00014005