A16612.22
A16612.36
A16612.22EA
62.1
EUR
InStock
A16612.22
A16612.36
1,2,3,4-Butantetracarbonsäure ≥98%
1,2,3,4-Butantetracarbonsäure
Purity: 98+%
CAS No.: 1703-58-8
Molecular Formula: C8H10O8
Formula Weight: 234.16
Melting Point: ca 190-195° dec.
Beilstein Reference: 1729167