A16652.06
A16652.14
A16652.06EA
45.3
EUR
InStock
A16652.06
A16652.14
2-Chlor-4-methoxyphenol ≥97%
2-Chlor-4-methoxyphenol
Purity: 97%
CAS No.: 18113-03-6
Molecular Formula: C7H7ClO2
Formula Weight: 158.59
Flash Point: 85°(185°F)
Odour: Phenol-like
Beilstein Reference: 1936412
MDL No.: MFCD00070773