A16786.06
A16786.14
A16786.06EA
77
EUR
InStock
A16786.06
A16786.14
2',3'-Difluoracetophenon ≥97%
2',3'-Difluoracetophenon
Purity: 97%
CAS No.: 18355-80-1
Molecular Formula: C8H6F2O
Formula Weight: 156.13
Refractive Index: 1.4890
Odour: Characteristic
Boiling Point: 84-85°/20mm
Beilstein Reference: 2963572
MDL No.: MFCD00061274