A16802.06
A16802.03
A16802.14
A16802.06EA
90.5
EUR
InStock
A16802.06
A16802.03
A16802.14
Acetylthiocholiniodid ≥98%
Acetylthiocholiniodid
Purity: 98%
CAS No.: 1866-15-5
Molecular Formula: CH3COSCH2CH2N(CH3)3I
Formula Weight: 289.18
Storage Temperature: 2°C to 8°C
Melting Point: 205-209°
Beilstein Reference: 3916578
UN No.: 2811
MDL No.: MFCD00011819