A17034.06
A17034.14
A17034.06EA
183
EUR
InStock
A17034.06
A17034.14
5-Hydroxy-1-tetralon ≥99%
5-Hydroxy-1-tetralon
Purity: 99%
CAS No.: 28315-93-7
Molecular Formula: C10H10O2
Formula Weight: 162.19
Melting Point: 209-211°
Beilstein Reference: 2437410