A17290.0B
A17290.18
A17290.30
A17290.0BEA
524
EUR
InStock
A17290.0B
A17290.18
A17290.30
2-Methylbenzimidazol ≥98%
2-Methylbenzimidazol
Purity: 98%
CAS No.: 615-15-6
Molecular Formula: C8H8N2
Formula Weight: 132.17
Melting Point: 174-178°
Beilstein Reference: 112264