A17338.0B
A17338.18
A17338.30
A17338.0BEA
636
EUR
InStock
A17338.0B
A17338.18
A17338.30
Methyltrifluoracetat ≥98%
Methyltrifluoracetat
Purity: 98+%
CAS No.: 431-47-0
Molecular Formula: CF3CO2CH3
Formula Weight: 128.05
Flash Point: -18°(-0.4°F)
Refractive Index: 1.2900
Odour: Pungent
Melting Point: -78°
Boiling Point: 43-44°
Beilstein Reference: 1756070
UN No.: 2924
MDL No.: MFCD00000417