A17692.03
A17692.06
A17692.03EA
64.6
EUR
InStock
A17692.03
A17692.06
8-Hydroxyjulolidine 97%
8-Hydroxyjulolidine
Purity: 97%
CAS No.: 41175-50-2
Molecular Formula: C12H15NO
Formula Weight: 189.26
Melting Point: 132-135°
Beilstein Reference: 1530423
MDL No.: MFCD00006918