A17743.09
A17743.18
A17743.30
A17743.09EA
33.8
EUR
InStock
A17743.09
A17743.18
A17743.30
m-Tolylessigsäure ≥99%
m-Tolylessigsäure
Purity: 99%
CAS No.: 621-36-3
Molecular Formula: C9H10O2
Formula Weight: 150.18
Melting Point: 62-66°
Beilstein Reference: 2043549