A17837.14
A17837.22
A17837.14EA
76.5
EUR
InStock
A17837.14
A17837.22
o-Tolidin ≥98%
o-Tolidin
Purity: 98%
CAS No.: 119-93-7
Molecular Formula: C14H16N2
Formula Weight: 212.30
Flash Point: 244°(471°F)
Melting Point: 128-132°
Boiling Point: 300-301°
Merck Reference: 14,9514
UN No.: 3077
MDL No.: MFCD00014773