A17867.14
A17867.22
A17867.36
A17867.14EA
64.5
EUR
InStock
A17867.14
A17867.22
A17867.36
2-Chlor-m-xylol ≥98%
2-Chlor-m-xylol
Purity: 98%
CAS No.: 6781-98-2
Molecular Formula: C8H9Cl
Formula Weight: 140.61
Flash Point: 63°(145°F)
Refractive Index: 1.5270
Melting Point: -36°
Boiling Point: 182-183°
Beilstein Reference: 2040954
MDL No.: MFCD00000565