A17998.14
A17998.22
A17998.14EA
79.1
EUR
InStock
A17998.14
A17998.22
4-Phenoxybenzoesäure ≥99%
4-Phenoxybenzoesäure
Purity: 99%
CAS No.: 2215-77-2
Molecular Formula: C13H10O3
Formula Weight: 214.22
Melting Point: 160-164°
Beilstein Reference: 2212463