A18115.14
A18115.22
A18115.36
A18115.14EA
41.6
EUR
InStock
A18115.14
A18115.22
A18115.36
4-Phenylbuttersäure ≥99%
4-Phenylbuttersäure
Purity: 99%
CAS No.: 1821-12-1
Molecular Formula: C6H5(CH2)3CO2H
Formula Weight: 164.20
Flash Point: 110°(230°F)
Melting Point: 49-53°
Boiling Point: 165°/10mm
Beilstein Reference: 638180
MDL No.: MFCD00004403