A18149.14
A18149.22
A18149.14EA
51.7
EUR
InStock
A18149.14
A18149.22
N,N-Diethyl-1,1,1-trimethylsilylamin 98%
N,N-Diethyl-1,1,1-trimethylsilylamin
Purity: 98%
CAS No.: 996-50-9
Molecular Formula: (CH3)3SiN(CH2CH3)2
Formula Weight: 145.32
Flash Point: 10°(50°F)
Refractive Index: 1.4110
Odour: Amine-like
Melting Point: -10°
Boiling Point: 125-126°
Beilstein Reference: 635718
UN No.: 2733
MDL No.: MFCD00009040