A18293.14
A18293.22
A18293.36
A18293.14EA
50
EUR
InStock
A18293.14
A18293.22
A18293.36
Rhodanin ≥98%
Rhodanin
Purity: 98+%
CAS No.: 141-84-4
Molecular Formula: C3H3NOS2
Formula Weight: 133.19
Odour: Odourless
Melting Point: 166-171°
Beilstein Reference: 110701
Merck Reference: 14,8184
MDL No.: MFCD00005488