A18740.06
A18740.14
A18740.06EA
46.9
EUR
InStock
A18740.06
A18740.14
3',4'-Dichlorpropiophenon ≥98%
3',4'-Dichlorpropiophenon
Purity: 98%
CAS No.: 6582-42-9
Molecular Formula: C9H8Cl2O
Formula Weight: 203.07
Flash Point: 110°(230°F)
Melting Point: 44-46°
Boiling Point: 136-140°/18mm
MDL No.: MFCD00000554