A18872.14
A18872.22
A18872.36
A18872.14EA
27
EUR
InStock
A18872.14
A18872.22
A18872.36
(±)-γ-Valerolacton ≥98%
(±)-γ-Valerolacton
Purity: 98+%
CAS No.: 108-29-2
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 81°(178°F)
Refractive Index: 1.4330
Odour: Pleasant
Melting Point: -31°
Boiling Point: 207-208°
Beilstein Reference: 80420
MDL No.: MFCD00005400