A18884.03
A18884.06
A18884.03EA
56.8
EUR
InStock
A18884.03
A18884.06
(4,5-Dimethoxy-2-nitrophenyl)essigsäure ≥97%
(4,5-Dimethoxy-2-nitrophenyl)essigsäure
Purity: 97%
CAS No.: 73357-18-3
Molecular Formula: C10H11NO6
Formula Weight: 241.20
Melting Point: 206-208°
MDL No.: MFCD00118466