A18932.06
A18932.14
A18932.06EA
35
EUR
InStock
A18932.06
A18932.14
Isobutylisothiocyanat ≥97%
Isobutylisothiocyanat
Purity: 97%
CAS No.: 591-82-2
Molecular Formula: (CH3)2CHCH2NCS
Formula Weight: 115.20
Flash Point: 24°(75°F)
Refractive Index: 1.4980
Odour: Pungent
Boiling Point: 160-162°
Beilstein Reference: 1740371
UN No.: 2924
MDL No.: MFCD00022062