A19010.14
A19010.03
A19010.06
A19010.14EA
354
EUR
InStock
A19010.14
A19010.03
A19010.06
Tetrahydro-4H-pyran-4-on ≥98%
Tetrahydro-4H-pyran-4-on
Purity: 98%
CAS No.: 29943-42-8
Molecular Formula: C5H8O2
Formula Weight: 100.12
Flash Point: 62°(144°F)
Refractive Index: 1.4530
Boiling Point: 165-167°
Beilstein Reference: 106463
MDL No.: MFCD00006581