A19016.22
A19016.36
A19016.22EA
36
EUR
InStock
A19016.22
A19016.36
(±)-β-Citronellol 95%
(±)-β-Citronellol
Purity: 95%
CAS No.: 106-22-9
Molecular Formula: C10H20O
Formula Weight: 156.27
Flash Point: 98°(208°F)
Refractive Index: 1.4575
Odour: Pleasant
Melting Point: -20°
Boiling Point: 221-222°
Beilstein Reference: 1721507
Merck Reference: 14,2330
UN No.: 3082
MDL No.: MFCD00002935