A19030.03
A19030.06
A19030.14
A19030.03EA
30
EUR
InStock
A19030.03
A19030.06
A19030.14
(R)-(-)-β-Aminophenethylalkohol ≥98%
(R)-(-)-β-Aminophenethylalkohol
Purity: 98%
CAS No.: 56613-80-0
Molecular Formula: C8H11NO
Formula Weight: 137.18
Melting Point: 76-78°
Beilstein Reference: 2935848