A19852.06
A19852.06EA
58.6
EUR
InStock
A19852.06
5-Chloro-2-mercaptobenzimidazole ≥98%
5-Chloro-2-mercaptobenzimidazole
Purity: 98%
CAS No.: 25369-78-2
Molecular Formula: C7H5ClN2S
Formula Weight: 184.64
Melting Point: 298-302°
UN No.: UN2811
MDL No.: MFCD01658762