A19878.22
A19878.14
A19878.22EA
151
EUR
InStock
A19878.22
A19878.14
3-Nonanon ≥98%
3-Nonanon
Purity: 98%
CAS No.: 925-78-0
Molecular Formula: C9H18O
Formula Weight: 142.24
Flash Point: 63°(145°F)
Refractive Index: 1.4200
Melting Point: -8°
Boiling Point: 187-188°
Beilstein Reference: 1700447
MDL No.: MFCD00009541