Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Azur A ≥70% (Farbstoffgehalt), Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

Achtung

M-Clarity™ Qualitätsstufe = MQ200

A6270-10G A6270-50G
A6270-10GEA 66.7 EUR InStock
A6270-10G A6270-50G
Azur A ≥70% (Farbstoffgehalt), Sigma-Aldrich®
Azur A
Azure A is a basic cationic dye. The combination of azure A and silver staining is used for the quantification of glycosaminoglycans in agarose gels. It is also used for the staining of chromosomes in the nucleus. It is a main component of the azure B/polychromed methylene blue – eosin stains family, also referred to as the Romanowsky stain.

Azure A chloride has been used for the staining of polyacrylamide gel to analyse the bovine lung heparin digestion by heparinase I. It has been used for the determination of the sulphated groups content in polysaccharides.

Achtung! Bitte beachten: Handle and store under argon. Glove bag item.


Formel: C₁₄H₁₄ClN₃S
MW: 291.797 g/mol
Melting Pt: 290 °C (Zers., Lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00012112
CAS-Nummer: 531-53-3
EINECS: 208-510-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=184c01e22be9943589aa6ffd4cf2c4961396b2bc&vid=184c01e22be9943589aa6ffd4cf2c4961396b2bc&prid=web&clid=SAPDAM
Order Now
Spezifikation Testergebnisse
Appearance (Colour) Blue to Very Dark Blue to Very Dark Green
Appearance (Form) Powder or Crystals
Infrared spectrum Conforms to Structure
Solubility (Colour) at 4 mg Plus 4 ml of Water Dark Blue to Very Dark Blue and Dark Purple to Very Dark Purple and Blue-Purple
Nitrogen 10.0 - 14.4 %
Dye Content ≥70 %

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge